missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-fluorophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65218.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499466

  • 2716.64 SEK / 1g


Estimated Shipment date
from Supplier 16-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27975737

Specifications

4-Chloro-N-(4-fluorophenyl)-3-sulfamoylbenzamide
MFCD27975737
Solid
1g
328.75
95%
Videos
SDS