missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-iodophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65362.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479586

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(4-iodophenyl)-3-sulfamoylbenzamide
436.65
MFCD28098789

Specifications

4-Chloro-N-(4-iodophenyl)-3-sulfamoylbenzamide
Light Sensitive
95%
MFCD28098789
1g
Solid
436.65
Videos
SDS