Learn More
Alfa Aesar™ 4-Chloro-N-(4-methoxybenzyl)-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65530.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449716
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 12-11-2018
Add to basket
Specifications
Chemical Identifiers
MFCD27975778
Specifications
4-Chloro-N-(4-methoxybenzyl)-3-sulfamoylbenzamide
MFCD27975778
Solid
1g
354.81
95%