Läs mer
4-Chloro-N-formylbenzamide, TRC
Produktkod. 30413250
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413250 250 mg
30423671 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR234590250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chloro-N-formylbenzamide
CAS 94670-39-0
Molekylformel C8 H6 Cl N O2
Kvantitet 250 mg
Synonym 4-Chloro-N-formylbenzamide
InChI formel InChI=1S/C8H6ClNO2/c9-7-3-1-6(2-4-7)8(12)10-5-11/h1-5H,(H,10,11,12)
LEDER Clc1ccc(cc1)C(=O)NC=O
IUPAC-namn 4-chloro-N-formylbenzamide
Molekylvikt (g/mol) 183.592
Formel vikt 183.009