Learn More
Alfa Aesar™ 4-Chloro-N-isopropyl-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65318.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15452519
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Chemical Identifiers
4-Chloro-N-isopropyl-3-sulfamoylbenzamide
276.74
MFCD21101912
Specifications
4-Chloro-N-isopropyl-3-sulfamoylbenzamide
Solid
276.74
1g
95%
MFCD21101912