Läs mer
4-Chloro Perazine Dihydrochloride, TRC
Produktkod. 30360600
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30360600 500 mg
1 options
Produktkod. 30360600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC374910500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chloro Perazine Dihydrochloride
CAS 1346602-34-3
Molekylformel C20H26Cl3N3S
Kvantitet 500 mg
Synonym 4-Chloro-10-[3-(4-methylpiperazin-1-yl)propyl]-10H-phenothiazine Dihydrochloride, Prochlorperazine Related Compound A
InChI formel InChI=1S/C20H24ClN3S.2ClH/c1-22-12-14-23(15-13-22)10-5-11-24-17-7-2-3-9-19(17)25-20-16(21)6-4-8-18(20)24;;/h2-4,6-9H,5,10-15H2,1H3;2*1H
LEDER ClC1=CC=CC2=C1SC3=CC=CC=C3N2CCCN4CCN(C)CC4.Cl.Cl
IUPAC-namn 4-chloro-10-(3-(4-methylpiperazin-1-yl)propyl)-10H-phenothiazine hydrogen chloride
Molekylvikt (g/mol) 446.86
Formel vikt 445.09