Läs mer
4-Chloro Trazodone Isomer, TRC
Produktkod. 30179889
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179889 250 mg
30180213 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179889 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC421650250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157072-19-0
Molekylformel C19 H22 Cl N5 O
Molekylvikt (g/mol) 371.8639
Synonym 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 2-[3-[4-(4-chlorophenyl)-1-piperazinyl]propyl]-, 2-[3-[4-(4-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, 4-Chlorotrazodone, 4-Chloro trazodone, 4-Chloro Trazodone Isomer
IUPAC-namn 2-[3-[4-(4-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one
LEDER Clc1ccc(cc1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2
Specifikationer
Kemiskt namn eller material 2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one
Kvantitet 250 mg
InChI formel InChI=1S/C19H22ClN5O/c20-16-5-7-17(8-6-16)23-14-12-22(13-15-23)9-3-11-25-19(26)24-10-2-1-4-18(24)21-25/h1-2,4-8,10H,3,9,11-15H2
Formel vikt 371.1513