Läs mer
4-Chlorobenzal Chloride, TRC
Produktkod. 30136609
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136609 10 g
30009525 5 g
30008320 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136609 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36415510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13940-94-8
Molekylformel C7H5Cl3
Molekylvikt (g/mol) 195.47
Synonym p,α,α-Trichlorotoluene, 1-Chloro-4-(dichloromethyl)benzene, 4-Chloro-1-(dichloromethyl)benzene, 4-Chlorobenzylidene Chloride, Dichloro(4-chlorophenyl)methane, p-(Dichloromethyl)benzyl Chloride, p-Chlorobenzal Chloride, p-Chlorobenzylidene Chloride, α,α,4-Trichlorotoluene, α,α,p-Trichlorotoluene
IUPAC-namn 1-chloro-4-(dichloromethyl)benzene
LEDER ClC1=CC=C(C=C1)C(Cl)Cl
Specifikationer
Kemiskt namn eller material 4-Chlorobenzal Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C7H5Cl3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,7H
Formel vikt 193.9457
Procent renhet >95%