Läs mer
4-Chlorobenzoic anhydride, TRC
Produktkod. 30365728
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365728 100 mg
30363314 250 mg
30357864 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365728 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC427630100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 790-41-0
Molekylformel C14H8Cl2O3
Molekylvikt (g/mol) 295.12
Synonym Benzoic acid, 4-chloro-, anhydride (9CI), Benzoic acid, p-chloro-, anhydride (6CI,7CI,8CI), 4-Chlorobenzoic acid anhydride, NSC 404992, p-Chlorobenzoic acid anhydride, p-Chlorobenzoic anhydride
IUPAC-namn (4-chlorobenzoyl) 4-chlorobenzoate
LEDER Clc1ccc(cc1)C(=O)OC(=O)c2ccc(Cl)cc2
Specifikationer
Kemiskt namn eller material 4-Chlorobenzoic anhydride
Kvantitet 100 mg
InChI formel InChI=1S/C14H8Cl2O3/c15-11-5-1-9(2-6-11)13(17)19-14(18)10-3-7-12(16)8-4-10/h1-8H
Formel vikt 293.985