Läs mer
4-Chlorobutyraldehyde Diethyl Acetal, TRC
Produktkod. 30361506
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30361506 5 g
1 options
Produktkod. 30361506 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3650515G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chlorobutyraldehyde Diethyl Acetal
CAS 6139-83-9
Molekylformel C8 H17 Cl O2
Kvantitet 5 g
Synonym Butane, 4-chloro-1,1-diethoxy-, Butyraldehyde, 4-chloro-, diethyl acetal (7CI,8CI), 4-Chloro-1,1-diethoxybutane, 4-Chlorobutanal diethyl acetal, 4-Chlorobutyraldehyde diethyl acetal, NSC 96467, γ-Chlorobutyraldehyde diethyl acetal
InChI formel InChI=1S/C8H17ClO2/c1-3-10-8(11-4-2)6-5-7-9/h8H,3-7H2,1-2H3
LEDER CCOC(CCCCl)OCC
IUPAC-namn 4-chloro-1,1-diethoxybutane
Molekylvikt (g/mol) 180.67
Formel vikt 180.0917