Läs mer
4-(Chloromethyl)-2-isopropyl]-4-thiazolyl, Hydrochloride Salt, TRC
Produktkod. 30353311
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30353311 50 mg
1 options
Produktkod. 30353311 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36919550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Chloromethyl)-2-isopropyl]-4-thiazolyl, Hydrochloride Sal
CAS 65386-28-9
Molekylformel C7 H10 Cl N S . Cl H
Kvantitet 50 mg
Synonym Thiazole, 4-(chloromethyl)-2-(1-methylethyl)-, hydrochloride (1:1), Thiazole, 4-(chloromethyl)-2-(1-methylethyl)-, hydrochloride (9CI), 4-(Chloromethyl)-2-isopropylthiazole hydrochloride
InChI formel InChI=1S/C7H10ClNS.ClH/c1-5(2)7-9-6(3-8)4-10-7;/h4-5H,3H2,1-2H3;1H
LEDER Cl.CC(C)c1nc(CCl)cs1
IUPAC-namn 4-(chloromethyl)-2-propan-2-yl-1,3-thiazole;hydrochloride
Molekylvikt (g/mol) 212.14
Formel vikt 210.9989