Läs mer
4-(Chloromethyl)-2-phenyl-1,3-oxazole, TRC
Produktkod. 30356369
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356369 50 mg
30363881 250 mg
30354403 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356369 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC65012550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30494-97-4
Molekylformel C10H8NOCl
Molekylvikt (g/mol) 193.63
Synonym 4-(Chloromethyl)-2-phenyloxazole, 2-Phenyl-4-chloromethyloxazole
IUPAC-namn 4-(chloromethyl)-2-phenyl-1,3-oxazole
LEDER ClCc1coc(n1)c2ccccc2
Specifikationer
Kemiskt namn eller material 4-(Chloromethyl)-2-phenyl-1,3-oxazole
Kvantitet 50 mg
InChI formel InChI=1S/C10H8ClNO/c11-6-9-7-13-10(12-9)8-4-2-1-3-5-8/h1-5,7H,6H2
Formel vikt 193.0294