missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-N-(3,5-difluorophenyl)benzamide, 97%, Thermo Scientific™

Catalog Number 15415906
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1094362-94-3
C14H10ClF2NO
281.687
MFCD12540820
AQOFMGRCNVYGFX-UHFFFAOYSA-N
4-chloromethyl-n-3,5-difluorophenyl benzamide
43156300
4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
C1=CC(=CC=C1CCl)C(=O)NC2=CC(=CC(=C2)F)F
This item is not returnable. View return policy
1094362-94-3
C14H10ClF2NO
281.687
MFCD12540820
AQOFMGRCNVYGFX-UHFFFAOYSA-N
4-chloromethyl-n-3,5-difluorophenyl benzamide
43156300
4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
C1=CC(=CC=C1CCl)C(=O)NC2=CC(=CC(=C2)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 281.69
Percent Purity 97%
Chemical Name or Material 4-Chloromethyl-N-(3,5-difluorophenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'