Läs mer
4-Chlorophenyl Benzoate, TRC
Produktkod. 30366432
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30366432 25 g
1 options
Produktkod. 30366432 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37518025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Chlorophenyl Benzoate
CAS 2005-08-5
Molekylformel C13H9ClO2
Kvantitet 25 g
Synonym Benzoic Acid, 4-Chlorophenyl Ester, Benzoic Acid, p-Chlorophenyl Ester, NSC 406605, p-Chlorophenol Benzoate
InChI formel InChI=1S/C13H9ClO2/c14-11-6-8-12(9-7-11)16-13(15)10-4-2-1-3-5-10/h1-9H
LEDER ClC1=CC=C(OC(C2=CC=CC=C2)=O)C=C1
IUPAC-namn 4-chlorophenyl benzoate
Molekylvikt (g/mol) 232.66
Formel vikt 232.03