Läs mer
4-Chlorophenylamine-13C6, TRC
Produktkod. 30365074
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365074 10 mg
30356467 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365074 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37750210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 89059-39-2
Molekylformel 13C6H6ClN
Molekylvikt (g/mol) 133.53
Synonym 4-Chloroaniline 13C6, Benzenamine-13C6, 4-chloro-, p-Aminochlorobenzene-13C6, p-Chloroaniline-13C6, 4-Chloro-1-aminobenzene-13C6, 4-Chloroaniline-13C6, 4-Chlorobenzenamine-13C6, 4-Amino-1-chlorobenzene-13C6, 4-Aminochlorobenzene-13C6, 1-Amino-4-chlorobenzene-13C6, NSC 36941-13C6
IUPAC-namn 4-chloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-amine
LEDER N[13c]1[13cH][13cH][13c](Cl)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material 4-Chlorophenylamine-13C6
Kvantitet 10 mg
InChI formel InChI=1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 133.039