missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyrazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7077825
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
15878-00-9
C3H3ClN2
102.52
BADSZRMNXWLUKO-UHFFFAOYSA-N
4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
27524
4-chloro-1H-pyrazole
C1=C(C=NN1)Cl
This item is not returnable. View return policy
15878-00-9
C3H3ClN2
102.52
BADSZRMNXWLUKO-UHFFFAOYSA-N
4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
27524
4-chloro-1H-pyrazole
C1=C(C=NN1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 71.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 102.52
Percent Purity 98%
Chemical Name or Material 4-Chloropyrazole
compliance-icons
Product Identifier
  • 4-Chloropyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'