missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals
217.75 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 228°C to 232°C |
Odor | Acrid |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 220.63 |
Percent Purity | 96% |
Chemical Name or Material | 4-(Chlorosulfonyl)benzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'