Läs mer
4-Cholesten-3-one-2,2,4,6,6-d5, TRC
Produktkod. 30363598
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363598 2 mg
30353923 1 mg
30363376 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4321022MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72560-60-2
Molekylformel C27 D5 H39 O
Molekylvikt (g/mol) 389.668
Synonym Cholest-4-en-3-one-2,2,4,6,6-d5 (9CI), 4-Cholesten-3-one-2,2,4,6,6-d5, 4-Cholesten-3-one-2,2,4,6,6-D5, δ4-Cholesten-3-one-d5, (17β)-17-Octylandrost-4-en-3-one-2,2,4,6,6-d5
IUPAC-namn (8S,9S,10R,13R,14S,17R)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
LEDER [2H]C1=C2C([2H])([2H])C[C@H]3[C@@H]4CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@@H]3[C@@]2(C)CC([2H])([2H])C1=O
Specifikationer
Kemiskt namn eller material 4-Cholesten-3-one-2,2,4,6,6-d5
Kvantitet 2 mg
InChI formel InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h17-19,22-25H,6-16H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1/i9D2,13D2,17D
Formel vikt 389.371