Läs mer
4-Cyano-4-[(ethylsulfanylthiocarbonyl)sulfanyl]pentanoic Acid, TRC
Produktkod. 30357833
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357833 50 mg
30357624 100 mg
30355683 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30357833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99857850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1137725-46-2
Molekylformel C9H13NO2S3
Molekylvikt (g/mol) 263.4
IUPAC-namn 4-cyano-4-ethylsulfanylcarbothioylsulfanylpentanoic acid
LEDER CCSC(=S)SC(C)(CCC(=O)O)C#N
Specifikationer
Kemiskt namn eller material 4-Cyano-4-[(ethylsulfanylthiocarbonyl)sulfanyl]pentanoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C9H13NO2S3/c1-3-14-8(13)15-9(2,6-10)5-4-7(11)12/h3-5H2,1-2H3,(H,11,12)
Formel vikt 263.0108