Läs mer
4-Cyano-4-(phenylcarbonothioylthio)pentanoic Acid, TRC
Produktkod. 30353900
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30353900 10 g
1 options
Produktkod. 30353900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98756010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Cyano-4-(phenylcarbonothioylthio)pentanoic Acid
CAS 201611-92-9
Molekylformel C13H13NO2S2
Kvantitet 10 g
Synonym 4-Cyano-4-[(phenylthioxomethyl)thio]pentanoic Acid, 1-(2-Carboxyethyl)-1-cyanoethyl benzodithioate, 4-(4-Cyanopentanoic Acid) Dithiobenzoate, 4-Cyano-4-(thiobenzoylthio)pentanoic acid, 4-Cyano-4-[(thiobenzoyl)sulfanyl]pentanoic acid, 6-Phenyl-6-thioxo-5-thia-4-cyano-4-methylhexanoic acid, Dithiobenzoic Acid 1-Cyano-1-methyl-3-carboxypropyl Ester
InChI formel InChI=1S/C13H13NO2S2/c1-13(9-14,8-7-11(15)16)18-12(17)10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,15,16)
LEDER CC(CCC(=O)O)(SC(=S)c1ccccc1)C#N
IUPAC-namn 4-(benzenecarbonothioylsulfanyl)-4-cyanopentanoic acid
Molekylvikt (g/mol) 279.38
Formel vikt 279.0388