missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanoindole, 99%, Thermo Scientific Chemicals

Catalog Number 15450059
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16136-52-0
C9H6N2
142.161
MFCD00152045
CEUFGDDOMXCXFW-UHFFFAOYSA-N
4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide
3817602
1H-indole-4-carbonitrile
C1=CC(=C2C=CNC2=C1)C#N
This item is not returnable. View return policy
16136-52-0
C9H6N2
142.161
MFCD00152045
CEUFGDDOMXCXFW-UHFFFAOYSA-N
4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide
3817602
1H-indole-4-carbonitrile
C1=CC(=C2C=CNC2=C1)C#N

4-Cyanoindole is used as a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanoindole is used as a synthetic intermediate.

Solubility
Soluble in water.

Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY
Melting Point 120°C to 125°C
Color Gray
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 142.16
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-Cyanoindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'