missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
Melting Point | ∼243°C (decomposition) |
Quantity | 5 g |
Beilstein | 3565486 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 169.61 |
Percent Purity | 97% |
Chemical Name or Material | 4-Cyanophenylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'