Läs mer
4-Defluoro-2-fluoro Haloperidol, TRC
Produktkod. 30011082
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011082 25 mg
30007211 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22878025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391052-53-1
Molekylformel C21 H23 Cl F N O2
Molekylvikt (g/mol) 375.86
Synonym 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(2-fluorophenyl)-, Haloperidol RC B (USP)
IUPAC-namn 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one
LEDER OC1(CCN(CCCC(=O)c2ccccc2F)CC1)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material 4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C21H23ClFNO2/c22-17-9-7-16(8-10-17)21(26)11-14-24(15-12-21)13-3-6-20(25)18-4-1-2-5-19(18)23/h1-2,4-5,7-10,26H,3,6,11-15H2
Formel vikt 375.1401
Procent renhet >95%