Läs mer
4-Dehydroxy-2-hydroxy Olsalazine, TRC
Produktkod. 30366818
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30366818 500 mg
1 options
Produktkod. 30366818 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229865500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,6'-Dihydroxy-3,3'-diazenediyldibenzoic Acid
CAS 752188-68-4
Molekylformel C14 H10 N2 O6
Kvantitet 500 mg
Synonym Benzoic acid, 3-[2-(3-carboxy-4-hydroxyphenyl)diazenyl]-2-hydroxy-, Benzoic acid, 3-[(3-carboxy-4-hydroxyphenyl)azo]-2-hydroxy- (9CI), 2,6'-Dihydroxy-3,3'-diazenediyldibenzoic acid, 4-Dehydroxy-2-hydroxy Olsalazine, 4-Dehydroxy-2-hydroxyolsalazine, Olsalazine Sodium Imp. B (EP)
InChI formel InChI=1S/C14H10N2O6/c17-11-5-4-7(6-9(11)14(21)22)15-16-10-3-1-2-8(12(10)18)13(19)20/h1-6,17-18H,(H,19,20)(H,21,22)
LEDER OC(=O)c1cccc(N=Nc2ccc(O)c(c2)C(=O)O)c1O
IUPAC-namn 3-[(E)-(3-carboxy-4-hydroxyphenyl)diazenyl]-2-hydroxybenzoic acid
Molekylvikt (g/mol) 302.24
Formel vikt 302.0539