Läs mer
4-Dehydroxy-4-chloro Olsalazine, TRC
Produktkod. 30354780
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30354780 100 mg
1 options
Produktkod. 30354780 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229830100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Chloro-6'-hydroxy-3,3'-diazenediyldibenzoic Acid
CAS 93964-55-7
Molekylformel C14 H9 Cl N2 O5
Kvantitet 100 mg
Synonym Benzoic acid, 5-[(3-carboxy-4-chlorophenyl)azo]-2-hydroxy- (9CI), 6-Chloro-6'-hydroxy-3,3'-diazenediyldibenzoic acid, 5-[(3-Carboxy-4-chlorophenyl)azo]salicylic acid, 4-Dehydroxy-4-chloro Olsalazine, 4-Dehydroxy-4-chlorolsalazine, Olsalazine Sodium Imp. D (EP)
InChI formel InChI=1S/C14H9ClN2O5/c15-11-3-1-7(5-9(11)13(19)20)16-17-8-2-4-12(18)10(6-8)14(21)22/h1-6,18H,(H,19,20)(H,21,22)
LEDER OC(=O)c1cc(ccc1O)N=Nc2ccc(Cl)c(c2)C(=O)O
IUPAC-namn 5-[(E)-(3-carboxy-4-chlorophenyl)diazenyl]-2-hydroxybenzoic acid
Molekylvikt (g/mol) 320.68
Formel vikt 320.02