Läs mer
4-Demethoxy-4-hydroxy Apixaban, TRC
Produktkod. 30138184
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138184 25 mg
30012251 10 mg
30004619 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23140525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 503612-76-8
Molekylformel C24 H23 N5 O4
Molekylvikt (g/mol) 445.47
Synonym 1H-Pyrazolo[3,4-c]pyridine-3-carboxamide, 4,5,6,7-tetrahydro-1-(4-hydroxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]- (9CI, ACI), 4,5,6,7-Tetrahydro-1-(4-hydroxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-1H-pyrazolo[3,4-c]pyridine-3-carboxamide (ACI), 1-(4-Hydroxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxamide, 1-(4-Hydroxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxamide
IUPAC-namn 1-(4-hydroxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxamide
LEDER NC(=O)c1nn(c2ccc(O)cc2)c3C(=O)N(CCc13)c4ccc(cc4)N5CCCCC5=O
Specifikationer
Kemiskt namn eller material 4-Demethoxy-4-hydroxy Apixaban
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C24H23N5O4/c25-23(32)21-19-12-14-28(16-6-4-15(5-7-16)27-13-2-1-3-20(27)31)24(33)22(19)29(26-21)17-8-10-18(30)11-9-17/h4-11,30H,1-3,12-14H2,(H2,25,32)
Formel vikt 445.175
Procent renhet >95%