Läs mer
4-Deoxo-4-hydroxy Spectinomycin Dihydrochloride, TRC
Produktkod. 30374679
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30374679 5 mg
1 options
Produktkod. 30374679 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6675505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4S)-Dihydrospectinomycin Dihydrochloride
CAS 56845-71-7
Molekylformel C14 H26 N2 O7 . 2 Cl H
Kvantitet 5 mg
Synonym 4-Deoxo-4-hydroxy Spectinomycin Dihydrochloride, 2H-Pyrano[2,3-b][1,4]benzodioxin-4,4a,7,9(10aH)-tetrol, octahydro-2-methyl-6,8-bis(methylamino)-, dihydrochloride, [2R-(2α,4α,4aβ,5aβ,6β,7β,8β,9α,9aα,10aβ)]- (9CI), (2R,4S,4aS,5aR,6S,7S,8R,9S,9aR,10aS)-2-Methyl-6,8-bis(methylamino)decahydro-2H-pyrano[2,3-
InChI formel InChI=1S/C14H26N2O7.2ClH/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;/h5-13,15-20H,4H2,1-3H3;2*1H/t5-,6+,7-,8+,9+,10+,11-,12-,13+,14+;;/m1../s1
LEDER Cl.Cl.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@@]3(O)[C@@H](O)C[C@@H](C)O[C@H]3O[C@@H]2[C@H]1O
Molekylvikt (g/mol) 407.287
Formel vikt 406.127