Läs mer
4-Deoxy Pyridoxine Hydrochloride, TRC
Produktkod. 17984665
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17984665 500 mg
17974645 250 mg
17933325 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17984665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD249930500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148-51-6
Molekylvikt (g/mol) 189.64
IUPAC-namn 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol;hydrochloride
Specifikationer
Kemiskt namn eller material 4-Deoxy Pyridoxine Hydrochloride
Anteckningar om renhetsgrad HPLC
Molekylformel C8H12ClNO2
Kvantitet 500 mg
Synonym 2,4-Dimethyl-3-hydroxy-5-hydroxymethylpyridine Hydrochloride, 4-Deoxypyridoxine Hydrochloride, 3-Hydroxy-5-hydroxymethyl-2,4-dimethylpyridine Hydrochloride, 4-Deoxypyridoxol Hydrochloride, 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride, 4-Desoxyadermin Hydrochloride, 4-Desoxypyridoxine Hydrochloride, 5-Hydroxymethyl-2,4-dimethylpyridin-3-ol Hydrochloride;
InChI formel InChI=1S/C8H11NO2.ClH/c1-5-7(4-10)3-9-6(2)8(5)11;/h3,10-11H,4H2,1-2H3;1H
LEDER CC1=C(C(=NC=C1CO)C)O.Cl
Formel vikt 189.056
Procent renhet >95%