Läs mer
4-Desacetamido-4-fluoro Andarine, TRC
Produktkod. 30133672
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133672 100 mg
30011564 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 401900-41-2
Molekylformel C17 H14 F4 N2 O5
Molekylvikt (g/mol) 402.3
Synonym Propanamide, 3-(4-fluorophenoxy)-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]-, (2S)- (9CI, ACI), (2S)-3-(4-Fluorophenoxy)-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide (ACI), ((S)-3-(4-Fluorophenoxy)-2-hydroxy-2-methyl-N-(4-nitro-3-trifluoromethylphenyl)propionamide, N-[4-Nitro-3-(trifluoromethyl)phenyl]-(2S)-3-(4-fluorophenoxy)-2-hydroxy-2-methylpropanamide, SARM S 1, 4-Desacetamido-4-fluoro Andarine
IUPAC-namn (2S)-3-(4-fluorophenoxy)-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
LEDER C[C@](O)(COc1ccc(F)cc1)C(=O)Nc2ccc(c(c2)C(F)(F)F)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 4-Desacetamido-4-fluoro Andarine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C17H14F4N2O5/c1-16(25,9-28-12-5-2-10(18)3-6-12)15(24)22-11-4-7-14(23(26)27)13(8-11)17(19,20)21/h2-8,25H,9H2,1H3,(H,22,24)/t16-/m0/s1
Formel vikt 402.0839
Procent renhet >95%