Läs mer
4-Desacetyl Vinblastine Methosulfate, TRC
Produktkod. 30353160
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30353160 25 mg
1 options
Produktkod. 30353160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28844025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Desacetyl Vinblastine Methosulfate
CAS 1322625-71-7
Molekylformel C44 H56 N4 O8 . C H4 O4 S
Kvantitet 25 mg
Synonym O4-Deacetyl-vincaleukoblastine Methosulfate, 4-Deacetylvinblastine Methosulfate, Deacetylvinblastine Methosulfate
InChI formel InChI=1S/C44H56N4O8.CH4O4S/c1-7-40(52)22-26-23-43(38(50)55-5,34-28(14-18-47(24-26)25-40)27-12-9-10-13-31(27)45-34)30-20-29-32(21-33(30)54-4)46(3)36-42(29)16-19-48-17-11-15-41(8-2,35(42)48)37(49)44(36,53)39(51)56-6;1-5-6(2,3)4/h9-13,15,20-21,26,35-37,45,49,52-53H,7-8,14,16-19,22-25H2,1-6H3;1H3,(H,2,3,4)/t26-,35-,36+,37+,40-,41+,42+,43-,44-;/m0./s1
LEDER CC[C@]1(O)C[C@@H]2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](O)[C@]8(CC)C=CCN9CC[C@]67[C@H]89)C(=O)OC)C1.COS(=O)(=O)O
Molekylvikt (g/mol) 881.04
Formel vikt 880.3928