Läs mer
4-Descarboxamido Rufinamide 4-Methyl Ester, TRC
Produktkod. 30365607
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30365607 5 mg
1 options
Produktkod. 30365607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2895555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 1-(2,6-difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate
CAS 217448-86-7
Molekylformel C11 H9 F2 N3 O2
Kvantitet 5 mg
Synonym Methyl 1-(2,6-difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate, Difluorobenzyl triazole ester, Triazole carboxylate derivative
InChI formel InChI=1S/C11H9F2N3O2/c1-18-11(17)10-6-16(15-14-10)5-7-8(12)3-2-4-9(7)13/h2-4,6H,5H2,1H3
LEDER COC(=O)c1cn(Cc2c(F)cccc2F)nn1
IUPAC-namn methyl 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxylate
Molekylvikt (g/mol) 253.2
Formel vikt 253.0663