Läs mer
4-Deschloro-4-bromo Dapagliflozin, TRC
Produktkod. 30010410
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010410 100 mg -
30004468 10 mg -
30004367 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD281960100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1807632-95-6
Molekylformel C21H25BrO6
Molekylvikt (g/mol) 453.32
Synonym (1S)-1,5-Anhydro-1-C-[4-bromo-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol, Dapagliflozin Impurity 3, (2S,3R,4R,5S,6R)-2-(4-bromo-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
LEDER CCOc1ccc(Cc2cc(ccc2Br)[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)cc1
Specifikationer
Kemiskt namn eller material 4-Deschloro-4-bromo Dapagliflozin
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C21H25BrO6/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21+/m1/s1
Formel vikt 452.0835
Procent renhet >95%