missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Diethylamino)salicylaldehyde, 98%, Thermo Scientific™
211.84 SEK - 695.91 SEK
Chemical Identifiers
CAS | 17754-90-4 |
---|---|
Molecular Formula | C11H15NO2 |
Molecular Weight (g/mol) | 193.24 |
InChI Key | XFVZSRRZZNLWBW-UHFFFAOYSA-N |
Synonym | 4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde |
PubChem CID | 87293 |
IUPAC Name | 4-(diethylamino)-2-hydroxybenzaldehyde |
SMILES | CCN(CC)C1=CC(=C(C=C1)C=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11935321
|
Thermo Scientific Acros
209480050 |
5g | Glass bottle |
211.84 SEK
5g |
Estimated Shipment: 08-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11391872
|
Thermo Scientific Acros
209480250 |
25g | Glass bottle |
695.91 SEK
25g |
Estimated Shipment: 08-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
17754-90-4 | |
193.24 | |
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde | |
4-(diethylamino)-2-hydroxybenzaldehyde |
C11H15NO2 | |
XFVZSRRZZNLWBW-UHFFFAOYSA-N | |
87293 | |
CCN(CC)C1=CC(=C(C=C1)C=O)O |
Specifications
4-(Diethylamino)salicylaldehyde | |
17754-90-4 | |
100.0 | |
60.0°C to 64.0°C | |
Authentic | |
14, 234 | |
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde | |
XFVZSRRZZNLWBW-UHFFFAOYSA-N | |
4-(diethylamino)-2-hydroxybenzaldehyde | |
87293 | |
Powder or Crystalline Powder |
98% | |
97.5 | |
Brown to Purple | |
97.5% min. (GC) | |
C11H15NO2 | |
Glass bottle | |
5g | |
CCN(CC)C1=CC(=C(C=C1)C=O)O | |
193.24 | |
193.24 | |
98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-(Diethylamino)salicylaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only