missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminophenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number 11308795
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
84381-54-4
C11H14N2S
206.31
MFCD00041094
PGWXUTHWTCRUAM-UHFFFAOYSA-N
4-diethylaminophenyl isothiocyanate, 4-diethylaminophenylisothiocyanate, 4-diethylamino benzenisothiocyanate, n,n-diethyl-4-isothiocyanato-aniline, #, benzenamine,n,n-diethyl-4-isothiocyanato, benzenamine, n,n-diethyl-4-isothiocyanato
145027
N,N-diethyl-4-isothiocyanatoaniline
CCN(CC)C1=CC=C(C=C1)N=C=S
This item is not returnable. View return policy
84381-54-4
C11H14N2S
206.31
MFCD00041094
PGWXUTHWTCRUAM-UHFFFAOYSA-N
4-diethylaminophenyl isothiocyanate, 4-diethylaminophenylisothiocyanate, 4-diethylamino benzenisothiocyanate, n,n-diethyl-4-isothiocyanato-aniline, #, benzenamine,n,n-diethyl-4-isothiocyanato, benzenamine, n,n-diethyl-4-isothiocyanato
145027
N,N-diethyl-4-isothiocyanatoaniline
CCN(CC)C1=CC=C(C=C1)N=C=S

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 33°C
Boiling Point 147°C to 148°C (1.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6685
Quantity 1 g
UN Number UN2923
Beilstein 3268361
Sensitivity Moisture sensitive
Formula Weight 206.31
Percent Purity 97%
Chemical Name or Material 4-Diethylaminophenyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'