missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzaldehyde, ≥99% (HPLC), Honeywell Fluka™

Catalog Number p-7120091
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1

for the determination of hydroxyproline,≥99.0% (HPLC)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 73°C to 75°C
Color White
Boiling Point 176°C to 177°C
Flash Point 164°C (327.2°F)
Ignition Residue ≤0.1% (as SO4)
Loss on Drying 0 to 0.5%
Packaging Glass Bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 50 g
UN Number NONH for all modes of transport
Beilstein 606802
Formula Weight 149.19g/mol
Percent Purity ≥99% (HPLC)
Chemical Name or Material 4-(Dimethylamino)benzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'