missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoic anhydride, 97%, Thermo Scientific Chemicals

Catalog Number p-7031430
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7474-31-9
C18H20N2O3
312.369
MFCD09757554
KGUATYWLOBCNMI-UHFFFAOYSA-N
4-dimethylaminobenzoic anhydride, 4-dimethylamino benzoyl 4-dimethylamino benzoate, acmc-20amjw, benzoic acid, 4-dimethylamino-, anhydride, 4-dimethylamino phenyl anhydride #, bis 4-dimethylamino benzoic anhydride, benzoic acid,4-dimethylamino-, anhydride with 4-dimethylamino benzoic acid
344732
[4-(dimethylamino)benzoyl] 4-(dimethylamino)benzoate
CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)N(C)C
This item is not returnable. View return policy
7474-31-9
C18H20N2O3
312.369
MFCD09757554
KGUATYWLOBCNMI-UHFFFAOYSA-N
4-dimethylaminobenzoic anhydride, 4-dimethylamino benzoyl 4-dimethylamino benzoate, acmc-20amjw, benzoic acid, 4-dimethylamino-, anhydride, 4-dimethylamino phenyl anhydride #, bis 4-dimethylamino benzoic anhydride, benzoic acid,4-dimethylamino-, anhydride with 4-dimethylamino benzoic acid
344732
[4-(dimethylamino)benzoyl] 4-(dimethylamino)benzoate
CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 149°C to 154°C
Quantity 1 g
Formula Weight 312.37
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminobenzoic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'