Läs mer
4-Dimethylaminobut-2-ynoic Acid Hydrochloride, TRC
Produktkod. 30372078
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30372078 500 mg
1 options
Produktkod. 30372078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD461620500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Dimethylaminobut-2-ynoic Acid Hydrochloride
CAS 118764-06-0
Molekylformel C6H9NO2 •HCl
Kvantitet 500 mg
Synonym Dimethylaminotetrolic acid, 4-(Dimethylamino)-2-butynoic Acid Hydrochloride, 4-(Dimethylamino)but-2-ynoic Acid Hydrochloride
InChI formel InChI=1S/C6H9NO2.ClH/c1-7(2)5-3-4-6(8)9;/h5H2,1-2H3,(H,8,9);1H
LEDER OC(C#CCN(C)C)=O.Cl
IUPAC-namn 4-(dimethylamino)but-2-ynoic acid hydrogen chloride
Molekylvikt (g/mol) 163.6
Formel vikt 163.04