Läs mer
4-Dimethylaminobutyraldehyde Diethyl Acetal, TRC
Produktkod. 30376415
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30376415 5 g
1 options
Produktkod. 30376415 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4610955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Dimethylaminobutyraldehyde Diethyl Acetal
CAS 1116-77-4
Molekylformel C11H23NO2
Kvantitet 5 g
Synonym 4,4-Diethoxy-N,N-dimethyl-1-butanamine, 1-(N,N-Dimethylamino)-4,4-diethoxybutane, 4,4-Diethoxy-N,N-dimethylbutanamine, 4,4-Diethoxy-N,N-dimethylbutylamine, 4-(Dimethylamino)butanal Diethyl Acetal, Zolmitriptan USP Related Compound H
InChI formel InChI=1S/C10H23NO2/c1-5-12-10(13-6-2)8-7-9-11(3)4/h10H,5-9H2,1-4H3
LEDER CCOC(CCCN(C)C)OCC
Molekylvikt (g/mol) 189.3
Formel vikt 189.1729