Läs mer
4-Dimethylaminosulfotoluidide, TRC
Produktkod. 30372994
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30372994 250 mg
1 options
Produktkod. 30372994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD462255250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DMST
CAS 66840-71-9
Anteckningar om renhetsgrad HPLC
Molekylformel C9 H14 N2 O2 S
Kvantitet 250 mg
Synonym N,N-dimethyl-N'-(4-methylphenyl)sulfamide, Sulfamide, N,N-dimethyl-N'-p-tolyl- (6CI), N,N-Dimethyl-N'-(4-methylphenyl)sulfamide, 4-Dimethylaminosulfotoluidide, DMST, N,N-dimethyl-N'-p-tolylsulfamide
InChI formel InChI=1S/C9H14N2O2S/c1-8-4-6-9(7-5-8)10-14(12,13)11(2)3/h4-7,10H,1-3H3
LEDER CN(C)S(=O)(=O)Nc1ccc(C)cc1
IUPAC-namn 1-(dimethylsulfamoylamino)-4-methylbenzene
Molekylvikt (g/mol) 214.28
Formel vikt 214.0776