missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylcarbamoyl)phenylboronic acid, 95%, Thermo Scientific™

Catalog Number 15204338
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
405520-68-5
C9H12BNO3
193.01
QJYYVSIRDJVQJW-UHFFFAOYSA-N
4-n,n-dimethylaminocarbonyl phenylboronic acid, 4-dimethylcarbamoyl phenyl boronic acid, 4-dimethylcarbamoyl phenylboronic acid, 4-n,n-dimethylcarbamoyl phenylboronic acid, 4-n,n-dimethylaminocarbonyl benzene boronic acid, 4-borono-n,n-dimethylbenzamide, boronic acid,b-4-dimethylamino carbonyl phenyl, 4-dimethylcarbamoyl benzeneboronic acid, boronic acid, 4-dimethylamino carbonyl phenyl
3503767
[4-(dimethylcarbamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C(=O)N(C)C)(O)O
This item is not returnable. View return policy
405520-68-5
C9H12BNO3
193.01
QJYYVSIRDJVQJW-UHFFFAOYSA-N
4-n,n-dimethylaminocarbonyl phenylboronic acid, 4-dimethylcarbamoyl phenyl boronic acid, 4-dimethylcarbamoyl phenylboronic acid, 4-n,n-dimethylcarbamoyl phenylboronic acid, 4-n,n-dimethylaminocarbonyl benzene boronic acid, 4-borono-n,n-dimethylbenzamide, boronic acid,b-4-dimethylamino carbonyl phenyl, 4-dimethylcarbamoyl benzeneboronic acid, boronic acid, 4-dimethylamino carbonyl phenyl
3503767
[4-(dimethylcarbamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C(=O)N(C)C)(O)O
Melting Point 144.0°C to 146.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 193.01
Percent Purity 95%
Chemical Name or Material 4-(Dimethylcarbamoyl)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(Dimethylcarbamoyl)phenylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'