Läs mer
4-Diphenylacetoxy-N-methylpiperidine methiodide, Thermo Scientific™
Produktkod. 15494699
Change view
Kvantitet:
25 mg
100 mg
Förpackningsstorlek:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15494699 25 mg 25mg
15420247 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15494699 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer J62298.MB
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1952-15-4
Molekylformel C21H26INO2
Molekylvikt (g/mol) 451.348
MDL-nummer MFCD00078564
InChI-nyckel WWJHRSCUAQPFQO-UHFFFAOYSA-M
Synonym 4-damp methiodide, 4-damp, 4-diphenylacetoxy-n-methylpiperidine methiodide, 4-damp iodide, 1,1-dimethyl-4-diphenylacetoxypiperidinium iodide, 4-diphenylacetyl oxy-1,1-dimethylpiperidinium iodide, piperidinium, 4-diphenylacetyl oxy-1,1-dimethyl-, iodide, 4-diphenylacetyl oxy-1,1-dimethyl-piperidinium iodide, 4-2,2-diphenylacetyl oxy-1,1-dimethylpiperidin-1-ium iodide, chembl76897
PubChem CID 3014059
ChEBI CHEBI:73340
IUPAC-namn (1,1-dimethylpiperidin-1-ium-4-yl) 2,2-diphenylacetate;iodide
LEDER C[N+]1(CCC(CC1)OC(=O)C(C2=CC=CC=C2)C3=CC=CC=C3)C.[I-]
Specifikationer
Kemiskt namn eller material 4-Diphenylacetoxy-N-methylpiperidine methiodide
Smältpunkt 211°C to 213°C
Färg White
Kvantitet 25 mg
Formel vikt 451.4
Fysisk form Powder