Läs mer
4-(Diphenylmethyl)-1-nitrosopiperidine, TRC
Produktkod. 30373021
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373021 100 mg
30379372 250 mg
30375327 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30373021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD940000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94067-39-7
Molekylformel C18H20N2O
Molekylvikt (g/mol) 280.37
Synonym Fexofenadine Impurity 25
LEDER O=NN1CCC(CC1)C(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material 4-(Diphenylmethyl)-1-nitrosopiperidine
Kvantitet 100 mg
InChI formel InChI=1S/C18H20N2O/c21-19-20-13-11-17(12-14-20)18(15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-10,17-18H,11-14H2
Formel vikt 280.1576