Läs mer
4-Dodecylresorcinol, TRC
Produktkod. 30375022
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375022 500 mg
30377321 1 g
30375344 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30375022 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD521185500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24305-56-4
Molekylformel C18 H30 O2
Molekylvikt (g/mol) 278.43
Synonym 4-Dodecyl-1,3-benzenediol, 4-Laurylresorcinol
IUPAC-namn 4-dodecylbenzene-1,3-diol
LEDER CCCCCCCCCCCCc1ccc(O)cc1O
Specifikationer
Kemiskt namn eller material 4-Dodecylresorcinol
Kvantitet 500 mg
InChI formel InChI=1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-16-13-14-17(19)15-18(16)20/h13-15,19-20H,2-12H2,1H3
Formel vikt 278.2246