Läs mer
4’-epi-Entecavir, TRC
Produktkod. 30179404
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30179404 25 mg -
30179228 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179404 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55893025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1367369-80-9
Molekylformel C12 H15 N5 O3
Molekylvikt (g/mol) 277.28
Synonym 2-Amino-9-[(1S,3R,4R)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-1,9-dihydro-6H-purin-6-one, 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4R)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, 4'-Epi-Entecavir, Entecavir Monohydrate Imp. D (EP)
IUPAC-namn 2-amino-9-[(1S,3R,4R)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-1H-purin-6-one
LEDER NC1=Nc2c(ncn2[C@H]3C[C@@H](O)[C@@H](CO)C3=C)C(=O)N1
Specifikationer
Kemiskt namn eller material 4'-epi-Entecavir
Kvantitet 25 mg
InChI formel InChI=1S/C12H15N5O3/c1-5-6(3-18)8(19)2-7(5)17-4-14-9-10(17)15-12(13)16-11(9)20/h4,6-8,18-19H,1-3H2,(H3,13,15,16,20)/t6-,7-,8+/m0/s1
Formel vikt 277.1175