Läs mer
4-Epidemeclocycline Hydrochloride, Mikromol™
Produktkod. 17982926
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
17982926 50 mg
1 options
Produktkod. 17982926 Leverantör Mikromol Leverantörsnummer MM0636.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Epidemeclocycline Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Epidemethylchlortetracycline Hydrochloride
CAS 179471-95-5
Analyt- eller komponentnamn 4-Epidemethylchlortetracycline Hydrochloride
Frakt skick Room Temperature
Molekylformel C21 H21 Cl N2 O8 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym [4R-(4íÅ1⁄2±,4aíÅ1⁄22,5aíÅ1⁄22,6íÅ1⁄2±,12aíÅ1⁄22)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, 4-Epidemeclocycline Hydrochloride, Chlortetracycline Hydrochloride Impurity E as Hydrochloride, Demeclocycline Hydrochloride Impurity B as Hydrochloride, Chlortetracycline Impurity E as Hydrochloride, Demeclocycline Impurity B as Hydrochloride
InChI formel InChI=1S/C21H21ClN2O8.ClH/c1-24(2)14-7-5-6-10(16(27)12-9(25)4-3-8(22)11(12)15(6)26)18(29)21(7,32)19(30)13(17(14)28)20(23)31;/h3-4,6-7,14-15,25-26,28-29,32H,5H2,1-2H3,(H2,23,31);1H/t6-,7-,14+,15-,21-;/m0./s1
LEDER Cl.CN(C)[C@@H]1[C@@H]2C[C@@H]3[C@H](O)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC-namn (4R,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
Molekylvikt (g/mol) 501.31
Kvantitet 50 mg
Formel vikt 500.0753 g/mol