missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog Number p-7081831
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=O)C=C1
This item is not returnable. View return policy
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=O)C=C1

4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY
Melting Point 13°C to 14°C
Density 1.08
Boiling Point 253°C to 255°C
Flash Point <110°C (230°F)
Refractive Index 1.559
Quantity 25 g
Beilstein 386863
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity ≥97%
Chemical Name or Material 4-Ethoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'