Läs mer
4-(Ethoxycarbonyl)benzoic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-(Ethoxycarbonyl)benzoic Acid |
| CAS | 713-57-5 |
| Molekylformel | C10H10O4 |
| Kvantitet | 50 mg |
| Synonym | 1,4-Benzenedicarboxylic acid, 1-ethyl ester (ACI), 1,4-Benzenedicarboxylic acid, monoethyl ester (9CI), Terephthalic acid, ethyl ester (6CI, 7CI), Terephthalic acid, monoethyl ester (8CI), 4-(Ethoxycarbonyl)benzoic acid, Ethyl 4-carboxybenzoate, Ethyl hydrogen terephthalate, Monoethyl terephthalate, Terephthalic acid, mono-ethyl ester |
| InChI formel | InChI=1S/C10H10O4/c1-2-14-10(13)8-5-3-7(4-6-8)9(11)12/h3-6H,2H2,1H3,(H,11,12) |
| LEDER | CCOC(=O)C1=CC=C(C=C1)C(=O)O |
| Molekylvikt (g/mol) | 194.18 |
| Formel vikt | 194.0579 |