Läs mer
4-Ethyl-5-fluoropyrimidine, TRC
Produktkod. 30009987
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009987 100 mg
30006508 1 g
30004577 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE918001100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137234-88-9
Molekylformel C6H7FN2
Molekylvikt (g/mol) 126.13
Synonym 4-Ethyl-5-fluoropyrimidine
IUPAC-namn 4-ethyl-5-fluoropyrimidine
LEDER CCc1ncncc1F
Specifikationer
Kemiskt namn eller material 4-Ethyl-5-fluoropyrimidine
Kvantitet 100 mg
InChI formel InChI=1S/C6H7FN2/c1-2-6-5(7)3-8-4-9-6/h3-4H,2H2,1H3
Formel vikt 126.0593