Learn More
4-Ethyl-N,N-diphenyl-1-naphthalenecarboxamide, TRC
Product Code. 30375071
Change view
Click to view available options
Kvantitet:
250 mg
Product Code. Kvantitet
30375071 250 mg
1 options
Product Code. 30375071 Supplier Toronto Research Chemicals Supplier No. TRCE917380250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-Ethyl-N,N-diphenyl-1-naphthalenecarboxamide
CAS 824430-38-8
Molekylformel C25H21NO
Kvantitet 250 mg
InChI formel InChI=1S/C25H21NO/c1-2-19-17-18-24(23-16-10-9-15-22(19)23)25(27)26(20-11-5-3-6-12-20)21-13-7-4-8-14-21/h3-18H,2H2,1H3
LEDER CCc1ccc(C(=O)N(c2ccccc2)c3ccccc3)c4ccccc14
IUPAC-namn 4-ethyl-N,N-diphenylnaphthalene-1-carboxamide
Molekylvikt (g/mol) 351.44
Formel vikt 351.1623