missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7080629
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
619-64-7
C9H10O2
150.177
MFCD00002570
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
12086
4-ethylbenzoic acid
CCC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
619-64-7
C9H10O2
150.177
MFCD00002570
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
12086
4-ethylbenzoic acid
CCC1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 111°C to 114°C
Quantity 10 g
Beilstein 2041840
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4-Ethylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'